Learn More
Waters Corp Waters Column, Spherisorb C1, 80Å, 3μm, 4.6 mm × 50 mm
Catalog No. 50784457 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb C1 columns are general purpose columns containing trimethylsilane bonded to silica, classified as USP L13.
  • General purpose column.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Maximum pressure of 6000 psi (415 Bar).
  • Not endcapped.
  • High silanol activity.
  • Spherical particle shape.
  • 3 μm particle size.
  • Parker-style endfitting type.
  • 80 Å pore size.
  • 220 surface area.
  • For HPLC systems.
  • L13 USP classification.
  • 4.6 mm inner diameter.
  • 50 mm length.
  • 2 % carbon load.
  • 1 unit per package.
Catalog No. 50-784-457 Supplier Waters Corp Supplier No. PSS832911
Spherisorb C1 columns are general purpose columns containing trimethylsilane bonded to silica, classified as USP L13.
  • General purpose column.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Maximum pressure of 6000 psi (415 Bar).
  • Not endcapped.
  • High silanol activity.
  • Spherical particle shape.
  • 3 μm particle size.
  • Parker-style endfitting type.
  • 80 Å pore size.
  • 220 surface area.
  • For HPLC systems.
  • L13 USP classification.
  • 4.6 mm inner diameter.
  • 50 mm length.
  • 2 % carbon load.
  • 1 unit per package.