Learn More
Waters Corp Spherisorb C1 Column, 80Å, 5 µm, 4.6 mm X 100 mm, 1/pk
Catalog No. 50784441 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb C1 columns are classified as USP L13 and are general purpose columns that contain trimethylsilane bonded to silica.
Catalog No. 50-784-441 Supplier Waters Corp Supplier No. PSS832612
Spherisorb C1 columns are classified as USP L13 and are general purpose columns that contain trimethylsilane bonded to silica.