Learn More
Waters Corp Waters Spherisorb Guard Cartridge, C1, 5 μm, 80 Å, 4.6 mm ID
Catalog No. 50784536 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb C1 columns are classified as USP L13 and are general purpose columns that contain trimethylsilane bonded to silica.
  • C1 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Spherical particle shape
  • 5 μm particle size
  • 80 Å pore size
  • Guard cartridge format
  • Designed for HPLC systems
  • 4.6 mm inner diameter
  • 30 mm length
  • 2% carbon load
  • Comes in a package of 3 units
Catalog No. 50-784-536 Supplier Waters Corp Supplier No. PSS839475
Spherisorb C1 columns are classified as USP L13 and are general purpose columns that contain trimethylsilane bonded to silica.
  • C1 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Spherical particle shape
  • 5 μm particle size
  • 80 Å pore size
  • Guard cartridge format
  • Designed for HPLC systems
  • 4.6 mm inner diameter
  • 30 mm length
  • 2% carbon load
  • Comes in a package of 3 units