Learn More
Waters Corp Waters Corporation Spherisorb Guard Cartridge, C1, 5 μm, 10 × 4.6 mm
Catalog No. 50784287 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Spherisorb Guard columns provide cost effective column protection for all Waters Spherisorb Analytical Columns.
  • Guard column design
  • C1 phase
  • 5 μm particle size
  • 10 × 4.6 mm dimensions
  • Pack of 3
  • Requires an In-line Guard Cartridge Holder
Catalog No. 50-784-287 Supplier Waters Corp Supplier No. PSS830076
Waters Spherisorb Guard columns provide cost effective column protection for all Waters Spherisorb Analytical Columns.
  • Guard column design
  • C1 phase
  • 5 μm particle size
  • 10 × 4.6 mm dimensions
  • Pack of 3
  • Requires an In-line Guard Cartridge Holder