Learn More
Waters Corp Waters Corporation Spherisorb C6 Column, 80Å, 5μm, 4.6 mm × 125 mm
Supplier:  Waters Corp PSS839848
Encompass_Preferred
Spherisorb C6 columns are general-purpose, silica-based reversed-phase columns that contain a linear-hexyl ligand. These USP L15 columns are used when a less retentive sorbent is required compared to ODS columns. The analytical column is produced under strict cGMP, ISO 9002 guidelines, ensuring top-notch lab equipment that generates reliable and consistent analysis results. The column offers excellent versatility in optimizing processes and reducing solvent use, and comes with crucial performance characteristics such as peak symmetry, lifespan, and column-to-column repeatability.
  • C6 chemistry
  • Reversed-phase separation mode
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • 5 μm particle size
  • 80 Å pore size
  • Suitable for HPLC systems
  • 4.6 mm inner diameter
  • 125 mm length
  • 5% carbon load
Catalog No. 50-784-635