Learn More
Waters Corp Waters Column, C6, Reversed Phase, 5 μm, 4.6 mm ID, 150 mm
Catalog No. 50784353 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb C6 columns are general purpose, silica-based reversed-phase columns with a linear-hexyl ligand, ideal for applications requiring less retention or rapid reversed-phase separation.
  • General-purpose, silica-based reversed-phase column.
  • Contains a linear-hexyl ligand for less retention than ODS columns.
  • Suitable for ion-pair work and rapid reversed-phase separations.
  • Features Parker-style column end fittings and high-quality bonded phases.
  • Durable and efficient for method optimization and solvent reduction.
  • Dimensions: 4.6 mm ID × 150 mm length.
  • Maximum pressure: 6000 psi (415 Bar); pH range: 2 to 8.
  • Manufactured under cGMP, ISO 9002 certified, and FDA registered for quality assurance.
Catalog No. 50-784-353 Supplier Waters Corp Supplier No. PSS831013
Spherisorb C6 columns are general purpose, silica-based reversed-phase columns with a linear-hexyl ligand, ideal for applications requiring less retention or rapid reversed-phase separation.
  • General-purpose, silica-based reversed-phase column.
  • Contains a linear-hexyl ligand for less retention than ODS columns.
  • Suitable for ion-pair work and rapid reversed-phase separations.
  • Features Parker-style column end fittings and high-quality bonded phases.
  • Durable and efficient for method optimization and solvent reduction.
  • Dimensions: 4.6 mm ID × 150 mm length.
  • Maximum pressure: 6000 psi (415 Bar); pH range: 2 to 8.
  • Manufactured under cGMP, ISO 9002 certified, and FDA registered for quality assurance.