Learn More
Waters Corp Waters Spherisorb C6 Column, 80Å, 5μm, 4 mm x 125 mm
Catalog No. 50784668 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • General purpose, silica-based reversed-phase columns.
  • Contain a linear-hexyl ligand.
  • Used when a less retentive sorbent is required compared to ODS columns.
Catalog No. 50-784-668 Supplier Waters Corp Supplier No. PSS845284
  • General purpose, silica-based reversed-phase columns.
  • Contain a linear-hexyl ligand.
  • Used when a less retentive sorbent is required compared to ODS columns.