Learn More
Waters Corp Spherisorb Column, C6, 80Å, 5 μm, 4.6 mm X 250 mm
Catalog No. 50784355 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C6 columns are general purpose, silica-based reversed-phase columns with a linear-hexyl ligand. They are USP L15 columns designed for applications requiring a less retentive sorbent compared to ODS columns, offering multi-purpose, long-lasting, and high-efficiency chromatographic performance. Meticulous packaging ensures maximum performance and reliable results.
  • General purpose, silica-based reversed-phase column with a linear-hexyl ligand.
  • Less retentive sorbent required for specific applications.
  • Multi-purpose, long-lasting, and high-efficiency.
  • Engineered to address process parameters for optimal column stability, efficiency, and symmetry.
  • Manufactured under strict cGMP and ISO 9002 compliance.
  • Ensures peak symmetry, column longevity, efficiency, and reproducibility.
Catalog No. 50-784-355 Supplier Waters Corp Supplier No. PSS831015
These C6 columns are general purpose, silica-based reversed-phase columns with a linear-hexyl ligand. They are USP L15 columns designed for applications requiring a less retentive sorbent compared to ODS columns, offering multi-purpose, long-lasting, and high-efficiency chromatographic performance. Meticulous packaging ensures maximum performance and reliable results.
  • General purpose, silica-based reversed-phase column with a linear-hexyl ligand.
  • Less retentive sorbent required for specific applications.
  • Multi-purpose, long-lasting, and high-efficiency.
  • Engineered to address process parameters for optimal column stability, efficiency, and symmetry.
  • Manufactured under strict cGMP and ISO 9002 compliance.
  • Ensures peak symmetry, column longevity, efficiency, and reproducibility.