Learn More
Waters Corp Spherisorb C8 Column, 80Å, 10 µm, 4 mm X 125 mm, 1/pk
Catalog No. 50784666 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb C8 columns are general purpose, silica-based reversed-phase columns that contain an Octyl ligand. These USP L7 columns are used when you require a less retentive sorbent compared to the Spheriorb ODS columns.
Catalog No. 50-784-666 Supplier Waters Corp Supplier No. PSS845282
Add to Cart
Add to Cart
Spherisorb C8 columns are general purpose, silica-based reversed-phase columns that contain an Octyl ligand. These USP L7 columns are used when you require a less retentive sorbent compared to the Spheriorb ODS columns.