Learn More
Waters Corp Spherisorb Column, C8, 3 μm, 4.6 mm × 50 mm
Catalog No. 50784412 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C8 columns are general purpose, silica-based reversed-phase columns that contain an Octyl ligand. They are USP L7 columns used when a less retentive sorbent compared to ODS columns is required.
  • General purpose, silica-based reversed-phase column.
  • Contains an Octyl ligand.
  • Has a USP L7 classification.
  • Offers less retention compared to ODS columns.
Catalog No. 50-784-412 Supplier Waters Corp Supplier No. PSS832211
These C8 columns are general purpose, silica-based reversed-phase columns that contain an Octyl ligand. They are USP L7 columns used when a less retentive sorbent compared to ODS columns is required.
  • General purpose, silica-based reversed-phase column.
  • Contains an Octyl ligand.
  • Has a USP L7 classification.
  • Offers less retention compared to ODS columns.