Learn More
Waters Corp Waters Spherisorb C8 Column, 80Å, 3 μm, 2 mm X 100 mm
Catalog No. 50784415 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb C8 columns are general purpose, silica-based reversed-phase columns that contain an Octyl ligand. These USP L7 columns are used when a less retentive sorbent compared to ODS columns is required.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range from 2 to 8
  • Maximum pressure of 415 bar (6000 psi)
  • Endcapped
  • Medium silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • Parker-style endfitting type
  • 80 Å pore size
  • Column format
  • 220 surface area
  • HPLC system compatible
  • USP L7 classification
  • 2 mm inner diameter
  • 100 mm length
  • 6 % carbon load
  • 1 unit per package
Catalog No. 50-784-415 Supplier Waters Corp Supplier No. PSS832222
Add to Cart
Add to Cart
Spherisorb C8 columns are general purpose, silica-based reversed-phase columns that contain an Octyl ligand. These USP L7 columns are used when a less retentive sorbent compared to ODS columns is required.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range from 2 to 8
  • Maximum pressure of 415 bar (6000 psi)
  • Endcapped
  • Medium silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • Parker-style endfitting type
  • 80 Å pore size
  • Column format
  • 220 surface area
  • HPLC system compatible
  • USP L7 classification
  • 2 mm inner diameter
  • 100 mm length
  • 6 % carbon load
  • 1 unit per package