Learn More
Waters Corp Waters Column, C8, 5 μm, 4.6 mm × 250 mm, 80 Å
Supplier:  Waters Corp PSS831815
Encompass_Preferred
These C8 columns are general-purpose, silica-based reversed-phase columns containing an Octyl ligand. As USP L7 columns, they are used when a less retentive sorbent is required. Featuring high-quality bonded phases for various separation selectivities and industry-standard Parker-style column end fittings, these analytical columns are durable and efficient, offering flexibility in optimizing methods and reducing solvent consumption.
  • General purpose, silica-based reversed-phase columns with an Octyl ligand.
  • USP L7 columns.
  • Manufactured under cGMP guidelines in an ISO 9002 certified plant.
  • Each column is individually tested with a test chromatogram supplied.
  • Durable and efficient, offering flexibility in optimizing methods and reducing solvent consumption.
  • Industry-standard Parker-style column end fittings.
Catalog No. 50-784-375