Learn More
Waters Corp Spherisorb Guard Cartridge, C8, 80Å, 5 μm, 4.6 mm X 30 mm
Catalog No. 50784534 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C8 guard cartridges are general-purpose, silica-based reversed-phase columns that feature an Octyl ligand. They are designed for applications requiring a less retentive sorbent compared to ODS columns.
  • General purpose, suitable for a wide range of chromatographic applications.
  • Silica-based reversed-phase, offering reliable performance for separations.
  • Octyl ligand provides specific selectivity for target analytes.
  • Less retentive sorbent, ideal for separations where reduced retention is desired.
  • Constructed with a rugged silica particle for extended column lifetime.
  • Each column is individually tested, and a test chromatogram is provided.
  • Utilizes reusable cartridge endfittings.
Catalog No. 50-784-534 Supplier Waters Corp Supplier No. PSS839473
These C8 guard cartridges are general-purpose, silica-based reversed-phase columns that feature an Octyl ligand. They are designed for applications requiring a less retentive sorbent compared to ODS columns.
  • General purpose, suitable for a wide range of chromatographic applications.
  • Silica-based reversed-phase, offering reliable performance for separations.
  • Octyl ligand provides specific selectivity for target analytes.
  • Less retentive sorbent, ideal for separations where reduced retention is desired.
  • Constructed with a rugged silica particle for extended column lifetime.
  • Each column is individually tested, and a test chromatogram is provided.
  • Utilizes reusable cartridge endfittings.