Learn More
Waters Corp Waters C8 Guard Cartridge, 80Å, 5 μm, 4.6 x 10 mm
Catalog No. 50784285 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge is a general purpose, silica-based reversed-phase column designed to extend the lifespan of chromatographic analytical columns. It effectively maintains UPLC, UHPLC, and HPLC separation efficacy by removing chemical and particle impurities from the mobile phase stream.
  • Increases chromatographic analytical column lifespan.
  • Maintains UPLC, UHPLC, and HPLC separation efficacy.
  • Removes chemical and particle impurities.
  • Compatible with C8 analytical columns (4.6 mm I.D., 5 μm sorbent particles).
  • Operates across a broad mobile phase pH range.
  • C8 ligand offers wide pH range, low pH stability, and minimal column bleed.
  • Manufactured with strict quality control and performance testing.
Catalog No. 50-784-285 Supplier Waters Corp Supplier No. PSS830074
This guard cartridge is a general purpose, silica-based reversed-phase column designed to extend the lifespan of chromatographic analytical columns. It effectively maintains UPLC, UHPLC, and HPLC separation efficacy by removing chemical and particle impurities from the mobile phase stream.
  • Increases chromatographic analytical column lifespan.
  • Maintains UPLC, UHPLC, and HPLC separation efficacy.
  • Removes chemical and particle impurities.
  • Compatible with C8 analytical columns (4.6 mm I.D., 5 μm sorbent particles).
  • Operates across a broad mobile phase pH range.
  • C8 ligand offers wide pH range, low pH stability, and minimal column bleed.
  • Manufactured with strict quality control and performance testing.