Learn More
Waters Corp Waters Column, Cyano (CN), 10 μm Particle, 4.6 mm ID, 250 mm Length
Catalog No. 50784480 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Spherisorb Cyano (CN) chemistry provides a general-purpose normal phase propyl cyano bonded ligand on a silica substrate. This column, conditioned with normal phase solvent, is ideal for separating hydrophobic compounds not eluted by standard C18/C8 columns. It offers rapid elution, selectivity for polar compounds, and excellent peak shape for strongly basic compounds.
  • General purpose normal phase propyl cyano bonded ligand on a silica substrate
  • Conditioned and shipped with normal phase solvent
  • Ensures excellent peak symmetry, efficiency, and enhanced column lifetime
  • Ideal for hydrophobic compounds not eluted by standard C18/C8 columns
  • Manufactured under cGMP, ISO 9002 certified, FDA registered, and individually tested with performance chromatogram on eCord™ chip
Catalog No. 50-784-480 Supplier Waters Corp Supplier No. PSS833515
The Spherisorb Cyano (CN) chemistry provides a general-purpose normal phase propyl cyano bonded ligand on a silica substrate. This column, conditioned with normal phase solvent, is ideal for separating hydrophobic compounds not eluted by standard C18/C8 columns. It offers rapid elution, selectivity for polar compounds, and excellent peak shape for strongly basic compounds.
  • General purpose normal phase propyl cyano bonded ligand on a silica substrate
  • Conditioned and shipped with normal phase solvent
  • Ensures excellent peak symmetry, efficiency, and enhanced column lifetime
  • Ideal for hydrophobic compounds not eluted by standard C18/C8 columns
  • Manufactured under cGMP, ISO 9002 certified, FDA registered, and individually tested with performance chromatogram on eCord™ chip