Learn More
Waters Corp Waters Spherisorb Cyano Column, 3 μm, 80 Å, 2 mm x 100 mm
Catalog No. 50784430 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Spherisorb CN chemistry is a general purpose normal phase propyl cyano bonded ligand to a silica substrate. This column is conditioned and shipped with normal phase solvent.
  • Cyano (CN) chemistry
  • Normal-phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8 pH
  • Maximum pressure of 6000 psi (415 Bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • Parker-style endfitting type
  • 80 Å pore size
  • 220 m²/g surface area
  • Designed for HPLC systems
  • L10 USP classification
  • 2 mm inner diameter
  • 100 mm length
  • 3% carbon load
  • 3.3 μmol/m² ligand density
Catalog No. 50-784-430 Supplier Waters Corp Supplier No. PSS832422
The Spherisorb CN chemistry is a general purpose normal phase propyl cyano bonded ligand to a silica substrate. This column is conditioned and shipped with normal phase solvent.
  • Cyano (CN) chemistry
  • Normal-phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8 pH
  • Maximum pressure of 6000 psi (415 Bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • Parker-style endfitting type
  • 80 Å pore size
  • 220 m²/g surface area
  • Designed for HPLC systems
  • L10 USP classification
  • 2 mm inner diameter
  • 100 mm length
  • 3% carbon load
  • 3.3 μmol/m² ligand density