Learn More
Waters Corp Waters Spherisorb Cyano Guard Cartridge, 5 μm, 80 Å
Catalog No. 50784537 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge utilizes Spherisorb CN chemistry, featuring a general-purpose normal phase propyl cyano bonded ligand to a silica substrate. It is designed to protect columns, extending their lifetime and ensuring optimal results.
  • General-purpose normal phase propyl cyano bonded ligand to a silica substrate.
  • Conditioned and shipped with normal phase solvent.
  • Versatile, reverse-phased, propyl cyano-bonded columns.
  • Provides best peak symmetry, column stability, and performance.
  • Protects columns for extended lifetime and optimal results.
  • Manufactured in specialized, tightly regulated chromatography chemistry factories.
  • Complies with cGMP and ISO 9002 standards.
  • Created using only the purest reagents with extensive testing for quality assurance.
Catalog No. 50-784-537 Supplier Waters Corp Supplier No. PSS839476
This guard cartridge utilizes Spherisorb CN chemistry, featuring a general-purpose normal phase propyl cyano bonded ligand to a silica substrate. It is designed to protect columns, extending their lifetime and ensuring optimal results.
  • General-purpose normal phase propyl cyano bonded ligand to a silica substrate.
  • Conditioned and shipped with normal phase solvent.
  • Versatile, reverse-phased, propyl cyano-bonded columns.
  • Provides best peak symmetry, column stability, and performance.
  • Protects columns for extended lifetime and optimal results.
  • Manufactured in specialized, tightly regulated chromatography chemistry factories.
  • Complies with cGMP and ISO 9002 standards.
  • Created using only the purest reagents with extensive testing for quality assurance.