Learn More
Waters Corp Waters Cyano (CN) Guard Cartridge, 80 Å, 5 μm, 4.6 mm X 10 mm
Catalog No. 50784288 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Cyano (CN) guard cartridges are propyl cyano bonded, general-purpose, reversed-phase columns. They offer excellent column stability, efficiency, and peak symmetry, contributing to high-performance laboratory analysis. Manufactured under stringent conditions, these cartridges adhere to cGMP and ISO 9002 standards, ensuring reliable equipment.
  • Propyl cyano bonded for general-purpose, reversed-phase separations.
  • Excellent column stability, efficiency, and peak symmetry.
  • Significant performance for quality lab analysis.
  • Designed for long column longevity and column-to-column repeatability.
  • Manufactured in dedicated and strictly controlled chemistry plants.
  • Adheres to cGMP and ISO 9002 standards.
  • Produced using pure reagents and extensive testing for quality assurance.
Catalog No. 50-784-288 Supplier Waters Corp Supplier No. PSS830077
These Cyano (CN) guard cartridges are propyl cyano bonded, general-purpose, reversed-phase columns. They offer excellent column stability, efficiency, and peak symmetry, contributing to high-performance laboratory analysis. Manufactured under stringent conditions, these cartridges adhere to cGMP and ISO 9002 standards, ensuring reliable equipment.
  • Propyl cyano bonded for general-purpose, reversed-phase separations.
  • Excellent column stability, efficiency, and peak symmetry.
  • Significant performance for quality lab analysis.
  • Designed for long column longevity and column-to-column repeatability.
  • Manufactured in dedicated and strictly controlled chemistry plants.
  • Adheres to cGMP and ISO 9002 standards.
  • Produced using pure reagents and extensive testing for quality assurance.