Learn More
Waters Corp Waters Column, Cyano (CN), Reversed Phase, 5 μm, 4.6 mm x 150 mm
Catalog No. 50784337 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Spherisorb CN chemistry features a general-purpose reversed-phase propyl cyano bonded ligand to a silica substrate. This column is conditioned and shipped with acetonitrile.
  • General-purpose reversed-phase propyl cyano bonded ligand to a silica substrate.
  • Conditioned and shipped with acetonitrile.
  • Ensure mobile phase compatibility before changing to a different mobile phase system.
  • Equilibrate the column using a minimum of 10 column volumes of the intended mobile phase.
  • Flush the column with five column volumes of a water or organic solvent mixture to avoid precipitating mobile-phase buffers.
  • Operates within an optimal pH range of 2 to 8.
Catalog No. 50-784-337 Supplier Waters Corp Supplier No. PSS830908
The Spherisorb CN chemistry features a general-purpose reversed-phase propyl cyano bonded ligand to a silica substrate. This column is conditioned and shipped with acetonitrile.
  • General-purpose reversed-phase propyl cyano bonded ligand to a silica substrate.
  • Conditioned and shipped with acetonitrile.
  • Ensure mobile phase compatibility before changing to a different mobile phase system.
  • Equilibrate the column using a minimum of 10 column volumes of the intended mobile phase.
  • Flush the column with five column volumes of a water or organic solvent mixture to avoid precipitating mobile-phase buffers.
  • Operates within an optimal pH range of 2 to 8.