Learn More
Waters Corp Waters Corporation Column, Amino (NH2), 5 μm, 4 mm x 250 mm
Supplier: Waters Corp PSS845301
Spherisorb NH2 (Amino) columns offer versatile performance for normal-phase, anion-exchange, and HILIC separations, utilizing a silica-based aminopropyl bonded sorbent.
- Silica-based aminopropyl bonded sorbent
- USP L8 classified
- Supports normal-phase, anion-exchange, HILIC
- Manufactured under cGMP, ISO 9002
- High efficiency, symmetry, longevity, reproducibility
- Wide application range (e.g., low molecular weight sugars)
- Parker-style end fittings
- Chemistry: Amino (NH2)
- Particle substrate: Silica
- pH range: 2 to 8 pH
- Max pressure: 6000 psi (415 Bar)
- Not endcapped
- Particle size: 5 μm
- Pore size: 80 Å
- Surface area: 220 m²/g
- Dimensions: 4 mm ID x 250 mm length
- Carbon load: 2%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.