Learn More
Waters Corp WATERS Spherisorb NH2 Column, 80Å, 3 μm, 4.6 mm x 50 mm
Catalog No. 50784419 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb NH2 (Amino) columns contain a silica-based aminopropyl bonded sorbent. The column is listed as USP L8 and is used for anion-exchange, normal-phase, and HILIC separations. These columns are described as general-purpose, long-lasting, and high-efficiency, intended to satisfy needs for improving processes and lowering solvent use.
  • Exceptional versatility in improving processes and lowering solvent use.
  • General-purpose, long-lasting, and high-efficiency.
  • Provides important performance qualities: column efficiency, peak symmetry, column longevity, and column-to-column reproducibility.
  • Offers excellent column stability and efficiency, along with the best peak symmetry.
  • Manufactured under strict cGMP, ISO 9002 conditions using only the purest reagents with multiple tests performed at each step to ensure high quality.
Catalog No. 50-784-419 Supplier Waters Corp Supplier No. PSS832311
Spherisorb NH2 (Amino) columns contain a silica-based aminopropyl bonded sorbent. The column is listed as USP L8 and is used for anion-exchange, normal-phase, and HILIC separations. These columns are described as general-purpose, long-lasting, and high-efficiency, intended to satisfy needs for improving processes and lowering solvent use.
  • Exceptional versatility in improving processes and lowering solvent use.
  • General-purpose, long-lasting, and high-efficiency.
  • Provides important performance qualities: column efficiency, peak symmetry, column longevity, and column-to-column reproducibility.
  • Offers excellent column stability and efficiency, along with the best peak symmetry.
  • Manufactured under strict cGMP, ISO 9002 conditions using only the purest reagents with multiple tests performed at each step to ensure high quality.