Learn More
Waters Corp Waters Column, Amino (NH2), 80Å, 5 μm, 4.6 mm X 50 mm
Catalog No. 50784359 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb NH2 (Amino) columns contain a silica-based aminopropyl bonded sorbent. The column is listed as USP L8 and is used for anion-exchange, normal-phase, and HILIC separations. These columns are designed with features to simplify method transfer and are suitable for amino acid-related separations.
  • Features MaxPeak HPS technology to reduce unwanted analyte-surface interactions.
  • Built on a rugged silica particle base for challenging separations and method transfer.
  • Listed as USP L8.
  • Mentioned in over 2,000 analytical abstracts, highlighting their essential role in the scientific community.
  • Compatible with VanGuard Column Protection to prevent contamination and extend column life.
  • Can function as a reverse-phase, normal-phase, or HILIC column.
Catalog No. 50-784-359 Supplier Waters Corp Supplier No. PSS831111
Add to Cart
Add to Cart
Spherisorb NH2 (Amino) columns contain a silica-based aminopropyl bonded sorbent. The column is listed as USP L8 and is used for anion-exchange, normal-phase, and HILIC separations. These columns are designed with features to simplify method transfer and are suitable for amino acid-related separations.
  • Features MaxPeak HPS technology to reduce unwanted analyte-surface interactions.
  • Built on a rugged silica particle base for challenging separations and method transfer.
  • Listed as USP L8.
  • Mentioned in over 2,000 analytical abstracts, highlighting their essential role in the scientific community.
  • Compatible with VanGuard Column Protection to prevent contamination and extend column life.
  • Can function as a reverse-phase, normal-phase, or HILIC column.