Learn More
Waters Corp Waters Corporation Spherisorb Amino (NH2) Guard Cartridge, 80Å, 5 μm, 4.6 mm × 30 mm, 3/pk
Catalog No. 50784539 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge is designed for high-performance liquid chromatography (HPLC) systems. It utilizes a silica-based aminopropyl bonded sorbent, classified as USP L8, for anion-exchange, normal-phase, and HILIC separations. It protects analytical columns by extracting particulate contaminants, extending their lifetime and ensuring data integrity.
  • Silica-based aminopropyl bonded sorbent.
  • USP L8 classification.
  • Supports anion-exchange, normal-phase, and HILIC separations.
  • Protects analytical columns from particulate contaminants.
  • Extends analytical column lifetime and ensures data integrity.
  • Withstands pressures up to 6,000 psi (415 Bar).
  • Matches analytical column dimensions for seamless protection.
  • Easily replaceable.
  • Requires a guard cartridge holder for installation.
Catalog No. 50-784-539 Supplier Waters Corp Supplier No. PSS839478
This guard cartridge is designed for high-performance liquid chromatography (HPLC) systems. It utilizes a silica-based aminopropyl bonded sorbent, classified as USP L8, for anion-exchange, normal-phase, and HILIC separations. It protects analytical columns by extracting particulate contaminants, extending their lifetime and ensuring data integrity.
  • Silica-based aminopropyl bonded sorbent.
  • USP L8 classification.
  • Supports anion-exchange, normal-phase, and HILIC separations.
  • Protects analytical columns from particulate contaminants.
  • Extends analytical column lifetime and ensures data integrity.
  • Withstands pressures up to 6,000 psi (415 Bar).
  • Matches analytical column dimensions for seamless protection.
  • Easily replaceable.
  • Requires a guard cartridge holder for installation.