Learn More
Waters Corp Spherisorb RP/Cyano (CN) Column, 80Å, 5 µm, 4.6 mm X 250 mm, 1/pk
Catalog No. 50784513 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb RP/CN columns contain a mixed-mode sorbent comprised of Nitrile (CN) and ODS (C18) ligands. The RP/CN sorbent provides alternative reversed-phase selectivity compared to the Spherisorb ODS sorbents.
Catalog No. 50-784-513 Supplier Waters Corp Supplier No. PSS837815
Spherisorb RP/CN columns contain a mixed-mode sorbent comprised of Nitrile (CN) and ODS (C18) ligands. The RP/CN sorbent provides alternative reversed-phase selectivity compared to the Spherisorb ODS sorbents.