Learn More
Waters Corp Waters Spherisorb ODS1 Column, 80Å, 5 μm, 4 x 150 mm
Catalog No. 50784632 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%.
  • General purpose, silica-based, reversed-phase C18 columns
  • Low ligand density
  • Specified carbon load of 6.2%
Catalog No. 50-784-632 Supplier Waters Corp Supplier No. PSS839845
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%.
  • General purpose, silica-based, reversed-phase C18 columns
  • Low ligand density
  • Specified carbon load of 6.2%