Learn More
Waters Corp Waters Corporation Column, C18, 5 μm, 4 mm X 250 mm, 80 Å
Catalog No. 50784698 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS1 columns are general-purpose, silica-based, reversed-phase C18 columns. These columns feature low ligand density with a specified carbon load of 6.2% and are designed to handle a wide range of applications.
  • General-purpose, silica-based, reversed-phase C18 column
  • Low ligand density with a specified carbon load of 6.2%
  • Designed to handle a wide range of applications
  • Packing process helps address peak symmetry, column efficiency, column lifetime, and column to column reproducibility
  • Each column is individually tested
  • A copy of the test chromatogram is supplied with each column
  • Recommended operating pH range is 2 to 8
Catalog No. 50-784-698 Supplier Waters Corp Supplier No. PSS845542
Spherisorb ODS1 columns are general-purpose, silica-based, reversed-phase C18 columns. These columns feature low ligand density with a specified carbon load of 6.2% and are designed to handle a wide range of applications.
  • General-purpose, silica-based, reversed-phase C18 column
  • Low ligand density with a specified carbon load of 6.2%
  • Designed to handle a wide range of applications
  • Packing process helps address peak symmetry, column efficiency, column lifetime, and column to column reproducibility
  • Each column is individually tested
  • A copy of the test chromatogram is supplied with each column
  • Recommended operating pH range is 2 to 8