Learn More
Waters Corp WATERS CORPORATION Spherisorb Column, 80Å, 5 μm, 4 mm X 125 mm
Catalog No. 50784697 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS1 columns are general-purpose, silica-based, reversed-phase C18 columns designed for precise liquid chromatography. They feature low ligand density and a specified carbon load of 6.2%, offering excellent separation of non-polar compounds for accurate and reproducible results. These columns are versatile for applications such as drug discovery, pharmaceutical analysis, environmental monitoring, and food analysis.
  • High-performance reversed-phase column for liquid chromatography
  • Silica-based with C18 chemistry and 6.2% carbon load
  • Excellent separation of non-polar compounds
  • Maximizes efficiency and extends equipment lifetime
  • Small internal diameter for small sample volumes
  • High efficiency and durability for consistent performance
  • 80 Å pore size
  • Maximum pressure of 6000 psi (415 bar)
  • Spherical particle shape with 220 m²/g surface area
Catalog No. 50-784-697 Supplier Waters Corp Supplier No. PSS845541
Spherisorb ODS1 columns are general-purpose, silica-based, reversed-phase C18 columns designed for precise liquid chromatography. They feature low ligand density and a specified carbon load of 6.2%, offering excellent separation of non-polar compounds for accurate and reproducible results. These columns are versatile for applications such as drug discovery, pharmaceutical analysis, environmental monitoring, and food analysis.
  • High-performance reversed-phase column for liquid chromatography
  • Silica-based with C18 chemistry and 6.2% carbon load
  • Excellent separation of non-polar compounds
  • Maximizes efficiency and extends equipment lifetime
  • Small internal diameter for small sample volumes
  • High efficiency and durability for consistent performance
  • 80 Å pore size
  • Maximum pressure of 6000 psi (415 bar)
  • Spherical particle shape with 220 m²/g surface area