Learn More
Waters Corp Waters Column, C18, 10 μm, 4 mm, 250 mm
Catalog No. 50784748 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general purpose, silica-based, reversed-phase C18 columns ensure optimal peak symmetry, efficiency, and stability with low ligand density and a 6.2% carbon load. They are manufactured in a cGMP, ISO 9001 certified facility.
  • General-purpose, silica-based, reversed-phase C18 column
  • Low ligand density with 6.2% carbon load for peak symmetry, efficiency, and repeatability
  • Packing materials designed for ACQUITY UPLC System
  • Each batch tested chromatographically with acidic, basic, and neutral analytes
  • Independently examined on eCord™ Intelligent Chip; Performance Chromatogram and Certificate of Batch Analysis provided
  • Effective across a 2 to 8 pH range
Catalog No. 50-784-748 Supplier Waters Corp Supplier No. PSS845619
These general purpose, silica-based, reversed-phase C18 columns ensure optimal peak symmetry, efficiency, and stability with low ligand density and a 6.2% carbon load. They are manufactured in a cGMP, ISO 9001 certified facility.
  • General-purpose, silica-based, reversed-phase C18 column
  • Low ligand density with 6.2% carbon load for peak symmetry, efficiency, and repeatability
  • Packing materials designed for ACQUITY UPLC System
  • Each batch tested chromatographically with acidic, basic, and neutral analytes
  • Independently examined on eCord™ Intelligent Chip; Performance Chromatogram and Certificate of Batch Analysis provided
  • Effective across a 2 to 8 pH range