Learn More
Waters Corp Spherisorb ODS1 Column, 80Å, 10 µm, 4.6 mm X 150 mm, 1/pk
Catalog No. 50784325 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%.
Catalog No. 50-784-325 Supplier Waters Corp Supplier No. PSS830713
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%.