Learn More
Waters Corp Waters Spherisorb ODS1 Column, 80Å, 3 μm, 4.6 mm × 125 mm
Catalog No. 50784474 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%. These columns are designed to handle a variety of applications.
  • Low ligand density packing for optimal performance.
  • Addresses crucial performance aspects including peak symmetry, column efficiency, column longevity, and column-to-column reproducibility.
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure reagents.
  • Independently tested, with a copy of the test chromatogram included with each column.
Catalog No. 50-784-474 Supplier Waters Corp Supplier No. PSS833416
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%. These columns are designed to handle a variety of applications.
  • Low ligand density packing for optimal performance.
  • Addresses crucial performance aspects including peak symmetry, column efficiency, column longevity, and column-to-column reproducibility.
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure reagents.
  • Independently tested, with a copy of the test chromatogram included with each column.