Learn More
Waters Corp Waters Guard Cartridge, 80Å, 5 μm, 4.6 mm x 10 mm
Catalog No. 50784284 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS1 guard cartridges are silica-based, reversed-phase C18 columns. They safeguard HPLC columns against contaminants, enhance separation efficiency, and deliver precise results with 80Å pore size and 5 μm particle size. Proprietary ODS1 chemistry ensures enhanced selectivity for polar compounds.
  • Safeguards against contaminants; enhances separation efficiency.
  • 80Å pore size and 5 μm particle size.
  • Proprietary ODS1 chemistry for enhanced selectivity.
  • Protects HPLC columns from solvent impurities, mobile phase additives, and sample matrix components.
  • Compatible with diverse HPLC systems.
  • 6% carbon load effectively removes organic impurities.
  • Easy installation and pre-conditioned for immediate usage.
  • Backed by a one-year warranty.
Catalog No. 50-784-284 Supplier Waters Corp Supplier No. PSS830073
Spherisorb ODS1 guard cartridges are silica-based, reversed-phase C18 columns. They safeguard HPLC columns against contaminants, enhance separation efficiency, and deliver precise results with 80Å pore size and 5 μm particle size. Proprietary ODS1 chemistry ensures enhanced selectivity for polar compounds.
  • Safeguards against contaminants; enhances separation efficiency.
  • 80Å pore size and 5 μm particle size.
  • Proprietary ODS1 chemistry for enhanced selectivity.
  • Protects HPLC columns from solvent impurities, mobile phase additives, and sample matrix components.
  • Compatible with diverse HPLC systems.
  • 6% carbon load effectively removes organic impurities.
  • Easy installation and pre-conditioned for immediate usage.
  • Backed by a one-year warranty.