Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Spherisorb ODS2 Column, 80 Å, 10 μm, 4 mm × 125 mm

Catalog No. 50784662 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns with intermediate ligand density and a specified carbon load of 11.5%. These durable, high-efficiency columns offer versatility for improving processes and lowering solvent use. Manufactured under strict cGMP and ISO 9002 conditions with ultra-pure reagents, they ensure maximum performance, stability, efficiency, peak symmetry, longevity, and column-to-column repeatability.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 pH to 8 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Bonding technology: ODS2
  • Silanol activity: High
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: 80 Å
  • Surface area: 220
  • System: HPLC
  • USP classification: L1
  • Carbon load: 12% (11.5% specified)

Catalog No. 50-784-662 Supplier Waters Corp Supplier No. PSS845278
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns with intermediate ligand density and a specified carbon load of 11.5%. These durable, high-efficiency columns offer versatility for improving processes and lowering solvent use. Manufactured under strict cGMP and ISO 9002 conditions with ultra-pure reagents, they ensure maximum performance, stability, efficiency, peak symmetry, longevity, and column-to-column repeatability.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 pH to 8 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Bonding technology: ODS2
  • Silanol activity: High
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: 80 Å
  • Surface area: 220
  • System: HPLC
  • USP classification: L1
  • Carbon load: 12% (11.5% specified)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.