Learn More
Waters Corp Waters Spherisorb ODS2 Column, 80Å Pore, 3 μm Particle, 4.6 mm x 125 mm
Catalog No. 50784407 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Spherisorb ODS2 Column is a general-purpose, silica-based, reversed-phase C18 column, featuring an ODS2 packing with intermediate ligand density and a specified carbon load of 11.5%. Designed for versatility, durability, and high-efficiency, it aims to improve processes and lower solvent use.
  • General purpose, silica-based, reversed-phase C18 column.
  • ODS2 packing with intermediate ligand density and 11.5% carbon load.
  • Designed for improved processes and reduced solvent use.
  • Durable and high-efficiency chromatographic lab equipment.
  • Manufactured with precise packing procedures for maximum performance.
  • Ensures optimal column stability, efficiency, and peak symmetry.
  • Produced under cGMP and ISO 9002 conditions using ultra-pure reagents.
  • Provides excellent column efficiency, peak symmetry, longevity, and column-to-column repeatability.
Catalog No. 50-784-407 Supplier Waters Corp Supplier No. PSS832116
The Spherisorb ODS2 Column is a general-purpose, silica-based, reversed-phase C18 column, featuring an ODS2 packing with intermediate ligand density and a specified carbon load of 11.5%. Designed for versatility, durability, and high-efficiency, it aims to improve processes and lower solvent use.
  • General purpose, silica-based, reversed-phase C18 column.
  • ODS2 packing with intermediate ligand density and 11.5% carbon load.
  • Designed for improved processes and reduced solvent use.
  • Durable and high-efficiency chromatographic lab equipment.
  • Manufactured with precise packing procedures for maximum performance.
  • Ensures optimal column stability, efficiency, and peak symmetry.
  • Produced under cGMP and ISO 9002 conditions using ultra-pure reagents.
  • Provides excellent column efficiency, peak symmetry, longevity, and column-to-column repeatability.