Learn More
Waters Corp Waters Spherisorb ODS2 Column, 5 μm, 4.6 mm X 200 mm
Catalog No. 50784386 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general purpose, silica-based, reversed-phase C18 columns feature intermediate ligand density and 11.5% carbon load. Manufacturing ensures consistent performance, optimal peak symmetry, column stability, and efficiency. This robust, high-efficiency column offers flexibility in technique optimization and lower solvent usage, designed for liquid chromatography under strict cGMP and ISO 9002 guidelines.
  • General purpose, silica-based, reversed-phase C18 column.
  • High-quality bonded phase provides distinct separation selectivities.
  • Functions across a 2–8 pH range.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Key performance characteristics include column efficiency, peak symmetry, longevity, and reproducibility.
Catalog No. 50-784-386 Supplier Waters Corp Supplier No. PSS831914
These general purpose, silica-based, reversed-phase C18 columns feature intermediate ligand density and 11.5% carbon load. Manufacturing ensures consistent performance, optimal peak symmetry, column stability, and efficiency. This robust, high-efficiency column offers flexibility in technique optimization and lower solvent usage, designed for liquid chromatography under strict cGMP and ISO 9002 guidelines.
  • General purpose, silica-based, reversed-phase C18 column.
  • High-quality bonded phase provides distinct separation selectivities.
  • Functions across a 2–8 pH range.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Key performance characteristics include column efficiency, peak symmetry, longevity, and reproducibility.