Learn More
Waters Corp Waters Spherisorb ODS2 Column, 80 Å, 5 μm, 4 mm x 250 mm
Catalog No. 50784661 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. With intermediate ligand density (11.5% carbon load), they suit diverse applications, offering method optimization flexibility and reduced solvent consumption. These durable, high-efficiency columns ensure optimal stability, peak symmetry, and reproducibility.
  • Silica-based, reversed-phase C18 column with 11.5% carbon load.
  • Suitable for a wide variety of chromatographic applications.
  • Offers flexibility in method optimization and reduces solvent consumption.
  • Manufactured under cGMP, ISO 9002 conditions for critical performance.
  • Ensures optimal column stability, high efficiency, and good peak symmetry.
Catalog No. 50-784-661 Supplier Waters Corp Supplier No. PSS845277
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. With intermediate ligand density (11.5% carbon load), they suit diverse applications, offering method optimization flexibility and reduced solvent consumption. These durable, high-efficiency columns ensure optimal stability, peak symmetry, and reproducibility.
  • Silica-based, reversed-phase C18 column with 11.5% carbon load.
  • Suitable for a wide variety of chromatographic applications.
  • Offers flexibility in method optimization and reduces solvent consumption.
  • Manufactured under cGMP, ISO 9002 conditions for critical performance.
  • Ensures optimal column stability, high efficiency, and good peak symmetry.