Learn More
Waters Corp Waters Spherisorb ODS2 Column, 3 μm, 4.6 mm × 250 mm, C18
Catalog No. 50784406 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns with intermediate ligand density and a specified carbon load of 11.5%. They are designed to ensure optimum performance, column stability, efficiency, and peak symmetry, and are manufactured under cGMP, ISO 9002 conditions.
  • General purpose, silica-based, reversed-phase C18 columns.
  • Intermediate ligand density with a specified carbon load of 11.5%.
  • Offers great flexibility in improving procedures and lowering solvent use.
  • Long-lasting and high-efficiency.
  • Ensures optimum column stability, efficiency, and peak symmetry.
  • Manufactured under the strictest cGMP, ISO 9002 conditions.
  • Offers critical performance features: column efficiency, peak symmetry, column lifetime, and column-to-column reproducibility.
Catalog No. 50-784-406 Supplier Waters Corp Supplier No. PSS832115
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns with intermediate ligand density and a specified carbon load of 11.5%. They are designed to ensure optimum performance, column stability, efficiency, and peak symmetry, and are manufactured under cGMP, ISO 9002 conditions.
  • General purpose, silica-based, reversed-phase C18 columns.
  • Intermediate ligand density with a specified carbon load of 11.5%.
  • Offers great flexibility in improving procedures and lowering solvent use.
  • Long-lasting and high-efficiency.
  • Ensures optimum column stability, efficiency, and peak symmetry.
  • Manufactured under the strictest cGMP, ISO 9002 conditions.
  • Offers critical performance features: column efficiency, peak symmetry, column lifetime, and column-to-column reproducibility.