Learn More
Waters Corp Waters Spherisorb ODS2 Column, 80Å, 3 μm, 4 mm x 60 mm, C18
Catalog No. 50784633 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general-purpose, silica-based, reversed-phase C18 columns. Featuring intermediate ligand density with a 11.5% carbon load, these high-performance columns offer a broad range of separation selectivities, optimal peak symmetry, stability, and efficiency. Produced under strict cGMP and ISO 9002 guidelines.
  • Offers flexibility in improving processes and reducing solvent use.
  • High-quality bonded phase enables broad separation selectivities.
  • Designed for peak efficiency by addressing all process parameters.
  • Meets quality criteria for liquid chromatography.
  • Produced under tight cGMP, ISO 9002 guidelines.
  • Provides excellent column efficiency, peak symmetry, longevity, and repeatability.
Catalog No. 50-784-633 Supplier Waters Corp Supplier No. PSS839846
Add to Cart
Add to Cart
Spherisorb ODS2 columns are general-purpose, silica-based, reversed-phase C18 columns. Featuring intermediate ligand density with a 11.5% carbon load, these high-performance columns offer a broad range of separation selectivities, optimal peak symmetry, stability, and efficiency. Produced under strict cGMP and ISO 9002 guidelines.
  • Offers flexibility in improving processes and reducing solvent use.
  • High-quality bonded phase enables broad separation selectivities.
  • Designed for peak efficiency by addressing all process parameters.
  • Meets quality criteria for liquid chromatography.
  • Produced under tight cGMP, ISO 9002 guidelines.
  • Provides excellent column efficiency, peak symmetry, longevity, and repeatability.