Learn More
Waters Corp Spherisorb ODS2 Column, 10 μm, 4.6 mm X 250 mm
Catalog No. 50784436 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%.
  • General purpose, silica-based, reversed-phase C18 column
  • Intermediate ligand density
  • Specified carbon load of 11.5%
Catalog No. 50-784-436 Supplier Waters Corp Supplier No. PSS832515
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%.
  • General purpose, silica-based, reversed-phase C18 column
  • Intermediate ligand density
  • Specified carbon load of 11.5%