Learn More
Waters Corp Waters Column, C18, 5 μm Particle, 4.6 mm ID, 50 mm Length
Supplier:  Waters Corp PSS831911
Encompass_Preferred
Spherisorb ODS2 columns are general-purpose, silica-based, reversed-phase C18 columns with intermediate ligand density and a specified carbon load of 11.5%. These durable, high-efficiency chromatographic columns offer flexibility in method optimization and solvent consumption reduction for various applications.
  • Manufactured under strict cGMP and ISO 9002 conditions
  • Provides excellent column efficiency, peak symmetry, and column-to-column reproducibility
  • High-quality bonded phase offers unique separation selectivities
  • Optimized packing processes ensure maximum performance, stability, and peak symmetry
Catalog No. 50-784-383