Learn More
Waters Corp Waters Column, Reversed Phase C18, 5 μm, 80 Å, 4.6 × 100 mm
Catalog No. 50784384 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%. These columns are made under stringent cGMP and ISO 9002 conditions in a dedicated chromatography chemistry production plant, ensuring consistent and trustworthy analysis findings. The high-quality bonded phase offers a wide range of separation selectivities.
  • Column efficiency
  • Peak symmetry
  • Column longevity
  • Column-to-column reproducibility
  • Suitable for liquid chromatography
  • Flexible for optimizing procedures
  • Lowers solvent use
  • Durable and high-efficiency chromatographic column
  • Offers a wide range of separation selectivities
Catalog No. 50-784-384 Supplier Waters Corp Supplier No. PSS831912
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%. These columns are made under stringent cGMP and ISO 9002 conditions in a dedicated chromatography chemistry production plant, ensuring consistent and trustworthy analysis findings. The high-quality bonded phase offers a wide range of separation selectivities.
  • Column efficiency
  • Peak symmetry
  • Column longevity
  • Column-to-column reproducibility
  • Suitable for liquid chromatography
  • Flexible for optimizing procedures
  • Lowers solvent use
  • Durable and high-efficiency chromatographic column
  • Offers a wide range of separation selectivities