Learn More
Waters Corp Spherisorb ODS2 Column, 80Å, 10 µm, 4.6 mm X 200 mm, 1/pk
Catalog No. 50784435 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%.
Catalog No. 50-784-435 Supplier Waters Corp Supplier No. PSS832514
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%.