Learn More
Waters Corp Spherisorb ODS2 Guard Cartridge, 80Å, 5 µm, 4.6 mm X 30 mm, 3/pk
Catalog No. 50784529 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%.
Catalog No. 50-784-529 Supplier Waters Corp Supplier No. PSS839458
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%.