Learn More
Waters Corp Waters Spherisorb ODSB Guard Cartridge, 80Å, 5 μm, 4.6 mm × 30 mm
Catalog No. 50784531 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These guard cartridges are general purpose, silica-based, reversed-phase C18 columns. The ODSB packing features intermediate ligand density with a specified carbon load of 11.5%.
  • General purpose, silica-based, reversed-phase C18 columns
  • Intermediate ligand density with a specified carbon load of 11.5%
Catalog No. 50-784-531 Supplier Waters Corp Supplier No. PSS839470
These guard cartridges are general purpose, silica-based, reversed-phase C18 columns. The ODSB packing features intermediate ligand density with a specified carbon load of 11.5%.
  • General purpose, silica-based, reversed-phase C18 columns
  • Intermediate ligand density with a specified carbon load of 11.5%