Learn More
Waters Corp Waters Spherisorb Phenyl Column, 80Å, 10 μm, 4 mm x 250 mm
Catalog No. 50784679 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb Phenyl columns contain a non-endcapped silica-based Phenyl phase that provides unique compound selectivity for reversed-phase applications.
  • Phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8 pH
  • Maximum pressure of 6000 psi (415 Bar)
  • Non-endcapped design
  • High silanol activity
  • Spherical particle shape
  • 10 μm particle size
  • Parker-style endfitting
  • 80 Å pore size
  • Column format
  • 220 surface area
  • Suitable for HPLC systems
  • USP classification L11
  • 4 mm inner diameter
  • 250 mm length
  • 3 % carbon load
  • 1 unit per package
Catalog No. 50-784-679 Supplier Waters Corp Supplier No. PSS845295
Spherisorb Phenyl columns contain a non-endcapped silica-based Phenyl phase that provides unique compound selectivity for reversed-phase applications.
  • Phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8 pH
  • Maximum pressure of 6000 psi (415 Bar)
  • Non-endcapped design
  • High silanol activity
  • Spherical particle shape
  • 10 μm particle size
  • Parker-style endfitting
  • 80 Å pore size
  • Column format
  • 220 surface area
  • Suitable for HPLC systems
  • USP classification L11
  • 4 mm inner diameter
  • 250 mm length
  • 3 % carbon load
  • 1 unit per package