Learn More
Waters Corp Waters Spherisorb Phenyl Column, 80Å, 5 μm, 4 mm × 250 mm
Catalog No. 50784677 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • This Spherisorb Phenyl Column features a non-endcapped silica-based Phenyl phase, offering unique compound selectivity for reversed-phase applications.
  • Non-endcapped silica-based Phenyl phase
  • Provides unique compound selectivity for reversed-phase applications
Catalog No. 50-784-677 Supplier Waters Corp Supplier No. PSS845293
  • This Spherisorb Phenyl Column features a non-endcapped silica-based Phenyl phase, offering unique compound selectivity for reversed-phase applications.
  • Non-endcapped silica-based Phenyl phase
  • Provides unique compound selectivity for reversed-phase applications