Learn More
Waters Corp Waters SAX Column, 80Å, 5 µm, 4 mm × 250 mm
Catalog No. 50784688 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb SAX columns feature a silica-based quaternary ammonium bonded sorbent, ideal for anion-exchange separations (USP L14). These general-purpose, durable, and high-efficiency columns offer flexibility in method optimization, reduced solvent consumption, and consistent performance, manufactured under cGMP, ISO 9002 conditions.
  • Silica-based quaternary ammonium bonded sorbent.
  • USP L14 classification for anion-exchange separations.
  • Flexible for method optimization and reduced solvent consumption.
  • Durable, general-purpose, and high-efficiency chromatographic column.
  • Manufactured under cGMP, ISO 9002 conditions.
  • Ensures critical performance: high efficiency, optimal stability, excellent peak symmetry, and extended column lifetime.
  • Addresses process parameters: slurry solvent, packing pressure, temperature, flow rate, and slurry volume.
Catalog No. 50-784-688 Supplier Waters Corp Supplier No. PSS845305
Spherisorb SAX columns feature a silica-based quaternary ammonium bonded sorbent, ideal for anion-exchange separations (USP L14). These general-purpose, durable, and high-efficiency columns offer flexibility in method optimization, reduced solvent consumption, and consistent performance, manufactured under cGMP, ISO 9002 conditions.
  • Silica-based quaternary ammonium bonded sorbent.
  • USP L14 classification for anion-exchange separations.
  • Flexible for method optimization and reduced solvent consumption.
  • Durable, general-purpose, and high-efficiency chromatographic column.
  • Manufactured under cGMP, ISO 9002 conditions.
  • Ensures critical performance: high efficiency, optimal stability, excellent peak symmetry, and extended column lifetime.
  • Addresses process parameters: slurry solvent, packing pressure, temperature, flow rate, and slurry volume.