Learn More
Waters Corp Spherisorb SAX Guard Cartridge, 80Å, 5 µm, 4.6 mm X 30 mm, 3/pk
Catalog No. 50784530 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb SAX columns contain a silica-based quaternary ammonium bonded sorbent. The column is listed as USP L14 and is used for anion-exchange separations.
Catalog No. 50-784-530 Supplier Waters Corp Supplier No. PSS839465
Spherisorb SAX columns contain a silica-based quaternary ammonium bonded sorbent. The column is listed as USP L14 and is used for anion-exchange separations.