Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Spherisorb SCX Column, 80Å, 5 μm, 4.6 mm X 150 mm

Catalog No. 50784503 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Spherisorb SCX columns contain an anionic silica-based sorbent for separating positively charged ionic species. This general-purpose, durable, and high-efficiency chromatographic column offers flexibility in optimizing methods while reducing solvent consumption. Manufactured under strict cGMP and ISO 9002 conditions using ultra-pure reagents, it ensures quality and reliability.

  • Cation-exchange chemistry
  • Ion-exchange separation mode
  • Silica particle substrate
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 Bar)
  • Not endcapped
  • SCX bonding technology
  • High silanol activity
  • Spherical particle shape
  • Particle size: 5 μm
  • Parker-style endfitting type
  • Pore size: 80 Å
  • Column format
  • Surface area: 220
  • HPLC system compatible
  • Inner diameter: 4.6 mm
  • Length: 150 mm
  • Carbon load: 4%

Catalog No. 50-784-503 Supplier Waters Corp Supplier No. PSS837513
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spherisorb SCX columns contain an anionic silica-based sorbent for separating positively charged ionic species. This general-purpose, durable, and high-efficiency chromatographic column offers flexibility in optimizing methods while reducing solvent consumption. Manufactured under strict cGMP and ISO 9002 conditions using ultra-pure reagents, it ensures quality and reliability.

  • Cation-exchange chemistry
  • Ion-exchange separation mode
  • Silica particle substrate
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 Bar)
  • Not endcapped
  • SCX bonding technology
  • High silanol activity
  • Spherical particle shape
  • Particle size: 5 μm
  • Parker-style endfitting type
  • Pore size: 80 Å
  • Column format
  • Surface area: 220
  • HPLC system compatible
  • Inner diameter: 4.6 mm
  • Length: 150 mm
  • Carbon load: 4%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.