Learn More
Waters Corp WATERS CORPORATION Column, 80Å, 5 μm, 4.6 mm, 250 mm
Catalog No. 50784504 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb SCX columns feature an anionic silica-based sorbent, providing strong cation-exchange for separating positively charged ionic species. These columns are engineered for high efficiency, optimal peak shape, and excellent reproducibility, ensuring a longer operational lifetime with a rugged silica particle design for enhanced durability.
  • Ion-exchange separation mode with silica particle substrate.
  • pH range of 2 to 8; maximum pressure of 6000 psi (415 bar).
  • Spherical particle (5 μm) with 80 Å pore size for HPLC systems.
  • 4.6 mm inner diameter; 250 mm length.
  • cGMP, ISO 9002 certified plant manufacturing; individually tested.
  • Enhances efficiency, peak shape, reproducibility, and lifetime.
Catalog No. 50-784-504 Supplier Waters Corp Supplier No. PSS837515
Spherisorb SCX columns feature an anionic silica-based sorbent, providing strong cation-exchange for separating positively charged ionic species. These columns are engineered for high efficiency, optimal peak shape, and excellent reproducibility, ensuring a longer operational lifetime with a rugged silica particle design for enhanced durability.
  • Ion-exchange separation mode with silica particle substrate.
  • pH range of 2 to 8; maximum pressure of 6000 psi (415 bar).
  • Spherical particle (5 μm) with 80 Å pore size for HPLC systems.
  • 4.6 mm inner diameter; 250 mm length.
  • cGMP, ISO 9002 certified plant manufacturing; individually tested.
  • Enhances efficiency, peak shape, reproducibility, and lifetime.