Learn More
Waters Corp Spherisorb SCX Column, 80Å, 10 µm, 4 mm X 125 mm, 1/pk
Catalog No. 50784693 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Spherisorb SCX columns contain an anionic silica-based sorbent. This strong cation- exchange sorbent is used to separate positively charged ionic species.
Catalog No. 50-784-693 Supplier Waters Corp Supplier No. PSS845310
Spherisorb SCX columns contain an anionic silica-based sorbent. This strong cation- exchange sorbent is used to separate positively charged ionic species.